missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ FluoroBlok™ Fibronectin Cell Culture Inserts
GREENER_CHOICE

Detect migrated cells below membrane using a fluorescence plate reader

Brand:  Corning™ 354597

Product Code. 10590442

  • £616.00 / Pack of 24
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Packaged 12 inserts in a 24 well plate, 2 plates per case

Corning BioCoat™ cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.

  • Available as convenient, individual inserts are packaged in ready-to-use 24-well plates and as a one-piece, automation friendly 24-Multiwell format (clear PET membrane only)
  • Combined benefits of Falcon™ inserts coated with HFN for enhanced cell attachment and growth
  • Individual inserts contains a 3.0 μm pore size PET or Corning™ FluoroBlok PET membrane

Storage: Store at 2-8°C, away from direct sunlight, do not freeze.; Shelf life: At least 3 months from date of shipment

Order Info

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4.2 cm2
3 μm
Promote attachment of BHK-21 Cells in Serum-Free Media
12/Pk.
24
PET
Plate
2° to 8°C
24/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.