Learn More
Corning™ BioCoat™ Gelatin Culture Dish
Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation
£87.87 - £301.19
Specifications
Bottom Shape | Round |
---|---|
Surface Treatment | Gelatin |
Diameter (Metric) | 100 mm |
Product Code | Brand | No. per Case | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Case | Price | Quantity & Availability | |||||
10135860
|
Corning™
354653 |
10/Cs. |
£87.87
Pack of 10 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
10574771
|
Corning™
356653 |
40 |
£301.19
Pack of 40 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
Description
Commonly used in the culture of normal and transfected cell types, including vascular endothelial (i.e., BME, BAEC), muscle, embryonic stem, HUVEC for E-Selectin expression and VEGF induction, and F9 teratocarcinoma cells for gene expression studies.
- Provide attachment and growth-promoting substrate for the culture of a variety of cells
- Lot-to-lot consistency results in reproducible experiments
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Round | |
Gelatin | |
100 mm |