Learn More
Corning™ BioCoat™ Gelatin Rectangular Canted Neck Culture Flasks w/Vented Cap
Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation
£86.20 - £510.00
Specifications
Capacity (English) | 8.45 oz. |
---|---|
Closure Material | Phenolic |
Capacity (Metric) | 250 mL |
For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
Neck Style | Canted |
Description
Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.
Applications include:
- Promotion of cell attachment and spreading
- Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
- Culture of HUVEC for E-Selectin7 expression and VEGF induction8
Source
Quality Control
- Tested for ability to promote proliferation of HUVECs
- Tested and found negative for bacteria and fungi
Storage
Specifications
8.45 oz. | |
250 mL | |
Canted | |
Gelatin |
Phenolic | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile |