missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Laminin Multiwell Plate

Promotion of cell attachment and spreading

Brand:  Corning™ 354412

Code : 67

Additional Details : Weight : 0.55000kg

Product Code. 10010781

  • £233.42 / Pack of 5
Estimated Shipment: 07-11-2024
to see stock.
Add to basket

Description

Description

Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.

Applications include:

  • Promotion of cell attachment and spreading
  • Induction of cell differentiation and neurite outgrowth
  • Increases proliferation of myoblasts
  • Studies of effects of laminin on cell behavior
  • Cell adhesion assays

Source

  • Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Quality Control

    • Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
    • Tested and found negative for bacteria and fungi
    • Laminin purity >90% by SDS-PAGE (contains entactin)

    Storage

  • 2°C to 8°C. Do not freeze.
  • Specifications

    Specifications

    4 x 6
    Yes
    Flat
    2 cm2
    Rectangular
    Cell Culture
    24
    Lid
    1/Pk.
    Mouse Laminin
    Clear
    Polystyrene
    Clear
    Polystyrene
    Non-sterile
    Multi-well Cell Culture Plate
    Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
    5/Cs.
    5/Cs.
    2°C to 8°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers