missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Laminin TC-treated Culture Dishes
GREENER_CHOICE

Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

Brand:  Corning™ 354405

 View more versions of this product

Product Code. 10448822

  • £231.82 / Pack of 20
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.

Applications include:

  • Promotion of cell attachment and spreading
  • Induction of cell differentiation and neurite outgrowth
  • Increases proliferation of myoblasts
  • Studies of effects of laminin on cell behavior
  • Cell adhesion assays

Source

  • Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Quality Control

    • Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
    • Tested and found negative for bacteria and fungi
    • Laminin purity >90% by SDS-PAGE (contains entactin)

    Storage

  • 2°C to 8°C. Do not freeze.
  • Order Info

    Bulk quantities available; please contact your Customer Service Representative.

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Round
    21.3 cm2
    Round
    20/Cs.
    Laminin
    Clear
    Polystyrene
    Cell Culture Dish
    5/Pk.
    60 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers