missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
£87.87 - £426.69
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Round |
Color | Clear |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
10399320
|
Corning™
354467 |
35 mm | 20/cs. |
£106.65
Pack of 20 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10478822
|
Corning™
356467 |
35 mm | 100/cs. |
£420.44
Pack of 100 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10003860
|
Corning™
354468 |
60 mm | 20/cs. |
£106.65
Pack of 20 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10145290
|
Corning™
354469 |
100 mm | 10/cs. |
£87.87
Pack of 10 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10204281
|
Corning™
356468 |
60 mm | 100/cs. |
£426.69
Pack of 100 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10632122
|
Corning™
356469 |
100 mm | 40/cs. |
£301.19
Pack of 40 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10307432
|
Corning™
354550 |
150 mm | 5/cs. |
£110.00
Pack of 5 |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
Description
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
- Coated with 0.2 micron filtered PDL in a cleanroom
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Round | |
Cell Culture Dish |
Closure Included | |
Round | |
Polystyrene | |
Poly-D-Lysine | |
Lid |