Learn More
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide
GREENER_CHOICE
Tested for the ability to promote neurite outgrowth with NG108 rat glioma/mouse neuroblastoma cells
Brand:  Corning™ 354688
Product Code. 10629842
  • £474.00 / Pack of 12
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning™ BioCoatTM 8-well culture slide with a uniform application of PDL/Laminin. Glass slide with polystyrene vessel, lid, and safety removal tool. 4/pack, 12/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: >= 3 months from date of shipment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Culture Slide
2° to 8°C
12/Cs.
Adhesion, growth and differentiation of neuronal and transfected cell lines
3/Pk.
8
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide > 8-well