Learn More
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells
Brand:  Corning™ 354518
Code : 67
Additional Details : Weight : 0.22000kg
 View more versions of this product
Product Code. 10574651
  • £106.65 / Pack of 20
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
Description
Description
Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.
  • Minimize assay failure due to cell detachment
  • Matrix components are applied as an even, optically clear coating
  • Precoated with Poly-L-Lysine to eliminate time-consuming preparation
  • Lot-to-lot consistency provides reproducible experiments
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
Round
Round
Cell Culture Dish
20/pk.
9.6 sq. cm
Poly-L-Lysine
20/cs.
35 mm