Learn More
Corning™ BioCoat™ Poly-L-Lysine Multiwell Plate
Attachment and spreading of a variety of cell types
Brand: Corning™ 356515
Code : 67
Additional Details : Weight : 3.28000kg
Description
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
Applications include:
- Attachment and spreading of a variety of cell types
- Cell differentiation and neurite outgrowth
- Attachment of fastidious transfected cell lines
- Support survival of primary neurons in culture
- Serum-free or reduced-serum culture
Source
- PDL, synthetic (MW 75-150 kD)
- PLL, synthetic (MW 30-70 kD)
Quality Control
- Tested for ability to promote firm attachment of RCG cells
- Tested and found negative for bacteria and fungi
Storage
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
2 x 3 | |
Yes | |
Flat | |
9.6 cm2 | |
Rectangular | |
ECM/Attachment Factors | |
6 | |
Lid | |
5/Pk. | |
50/Cs. | |
4° to 30°C |
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Non-sterile | |
Multi-well Cell Culture Plate | |
Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases | |
50/Cs. | |
10 inner packs of 5 | |
Poly-L-Lysine |