missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Black 384-Well Polypropylene Assay Plate

Product Code. 10400672 Shop All Corning Products
Click to view available options
Lid:
With Lid
Without Lid
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 10400672

Brand: Corning™ 3658

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lower compound sticking and higher solvent resistance than polystyrene plates

  • Ideal for fluorescence assays and for compound storage
  • Round-bottom wells
  • Untreated

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Color Black
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 0.015 to 0.07 mL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) For fluorescence assays and compound storage
Quantity 100/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.