missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Black Universal Microplate Lids

Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ Black Universal Microplate Lids are suitable for fluorescent and other light-sensitive assays.
Brand: Corning™ 3935
91.22 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Black | |
Sterile | |
25/Pk. | |
Plate Lid |
Polystyrene | |
50/Cs. | |
For fluorescent and other light-sensitive assays |
Product Suggestions
Customers who viewed this item also viewed
COR001 Lifescience Guid
File size: 7 MB
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
16Hours
21Minutes
37Seconds
34New Offer Save Up to 92%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Black Universal Microplate Lids > Black Universal Lid with Corner Notch; 50/Cs
Spot an opportunity for improvement?Share a Content Correction