missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Black and White 96-Well Polypropylene Assay Plates

Lower compound sticking and higher solvent resistance than polystyrene plates
£372.00 - £483.21
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
17710698
|
Corning™
3355 |
White |
£372.00
Pack of 100 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
10285421
![]() |
Corning™
3356 |
Black |
£483.21
Pack of 100 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
Description
- Ideal for fluorescence and luminescence assays and for compound storage
- Round-bottom wells
- Untreated surface
Specifications
Standard | |
Polypropylene | |
75 to 200 μL | |
Non-treated |
8 x 12 | |
96 | |
Non-sterile | |
Ideal for fluorescence and luminescence assays and for compound storage |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Black and White 96-Well Polypropylene Assay Plates