Learn More
Corning™ Breathable Sealing Tape for Microplates
Fits 96- and 384-well microplates, polypropylene blocks
Brand: Corning™ 3345
This item is not returnable.
View return policy
Description
- Easy application and removal for short- and long-term storage
- Provides tight seal to minimize evaporation and condensation
- Breathable sealing tape allows gas exchange across the surface
- Sealing tape 3345 is not transparent
Other Available Options:
- Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
- Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
Specifications
Sterile | |
50/Pk. | |
Yes |
500/Cs. | |
Breathable Sealing Tape |