missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Calcein AM Fluorescent Dye

Reliable and consistent dye used for the detection of live cells and key functional activities in variety of cell-based assays

£285.00 - £304.00

Specifications

Content And Storage -20°C
For Use With (Equipment) Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10520286
View Documents
Corning™
354216
500 μg
£285.00
500µg
Please to purchase this item. Need a web account? Register with us today!
10530286
View Documents
Corning™
354217
1 mg
£304.00
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Specifications

Specifications

-20°C
Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells
Videos
SDS
Documents

Documents

Product Certifications

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.