Learn More
Corning™ Carbo-BIND Microplates
Surface has hydrazide groups covalently coupled to carbohydrate groups
Brand: Corning™ 2507
Code : 66
Additional Details : Weight : 0.02239kg
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
50/Cs. | |
Clear |
Without Lid | |
Carbo-BIND | |
Polystyrene | |
75 to 200 μL | |
Carbo-Bind | |
Ideal for assays requiring site-directed orientation of a biomolecule (oxidized antibodies, carbohydrates and glycosylated proteins) while maintaining enzymatic or immunological activity | |
1/Pk. |