missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell-Tak Cell and Tissue Adhesive

Product Code. p-90828 Shop All Corning Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 11593550

Brand: Corning™ 354242

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stable for at least three months at 2° to 8°C. Do not freeze.
Description Molecular weight: 110 to 114kd
Concentration 1 to 5μg/ cm2 of growth surface depending on cell type
Form Liquid
Product Type Cell and Tissue Adhesive
Formulation As liquid in 5% acetic acid
Quantity 10 mg
Source Polyphenolic Proteins secreted by Mytilus edulis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.