missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ Surface Culture Dishes
GREENER_CHOICE

Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.

Brand:  Corning™ 3295

 View more versions of this product

Product Code. 10665893

  • £216.00 / Pack of 126
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
  • Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
  • 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
  • 245 mm square dishes offer 500 cm2 growth surface
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Manufactured from optically clear virgin polystyrene
  • Sterile
  • Nonpyrogenic


Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.

Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
6.3 mL (Well)
21 cm2
Polystyrene
Sterile
Cell Culture Petri Dish
126/Cs.
15 mm
Yes
Round
Clear
Single-use
Round
CellBIND
Lid
7/Pk.
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.