Learn More
Corning™ CellCube™ Tubing Adaptor
A new closed system offering designed to help reduce the risk of adventitious contamination
Brand:  Corning™ 3237
Additional Details : Weight : 0.85500kg
 View more versions of this product
Product Code. 17259109
  • £307.99 / Pack of 4
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Closed System Cell Cube
  • CellCube 10, 25, 100 modules, circulation loops, and connectors
  • Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
Specifications
9.53 mm
4/Cs.
AseptiQuik™ S Connected to a Female MPC
Thermoelastic Tubing, Aseptic Connector to Female MPC
1/Pk.