missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Antibiotic-Antimycotic Solution

Packaged in plastic bottles

Brand:  Corning™ 30-004-CI

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 15343671

  • £331.00 / Pack of 6
In Stock
to see stock.

Description

Description

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

Specifications

Specifications

-20°C
Solution
Sterile
6 x 100 mL
10,000 IU Penicillin, 10,000μg/mL Streptomycin and 25μg/mL Amphotericin B
Antibiotic-Antimycotic
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers