missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 10X
With low endotoxin content
£197.26
Specifications
Content And Storage | 2° to 30°C |
---|---|
Concentration | 10 X |
Form | Liquid |
Product Type | Dulbecco's Phosphate Buffered Saline |
Quantity | 6 x 500 mL |
Product Code | Brand | Form | Quantity | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Product Type | Price | Quantity & Availability | |||
15333571
|
Corning™
20-030-CV |
Liquid | 6 x 500 mL | Dulbecco's Phosphate Buffered Saline |
£197.26
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.Specifications
2° to 30°C | |
Liquid | |
6 x 500 mL |
10 X | |
Dulbecco's Phosphate Buffered Saline |