Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
With low endotoxin content.
£80.90 - £169.33
Product Code | Brand | Product Type | Form | Quantity | Additive | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Form | Quantity | Additive | Price | Quantity & Availability | ||
15333631
|
Corning™
21-030-CM |
DPBS | Liquid | 6 x 1000 mL | Calcium and Magnesium |
£169.33
Pack of 6 |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15393571
|
Corning™
21-030-CV |
DPBS | Liquid | 6 x 500 mL | Calcium and Magnesium |
£80.90
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15343631
|
Corning™
21-031-CM |
DPBS | Liquid | 6 x 1000 mL | No Calcium, No Magnesium |
£162.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15303581
|
Corning™
21-031-CV |
DPBS | Liquid | 6 x 500 mL | No Calcium, No Magnesium |
£86.75
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.Specifications
1 X | |
DPBS |