missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Media: Cell Culture Supplements
BIOPHARMA

Product Code. p-3202715
Click to view available options
pH:
5.2 to 0.1
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15313721

Brand: Corning™ 46033CI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a nutrient source in cell culture media. Cell Culture Media: Cell Culture Supplements are produced under the highest industry standards to ensure superior results.

  • Certificate of Analysis available
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Sodium Acetate
Sterility Sterile-filtered
pH 5.2 to 0.1
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.