missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Cell Culture Media - Dulbecco's Modification of Eagle's Medium (DMEM)
Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over the original formulation. Corning™ Cell Culture Media - Dulbecco's Modification of Eagle's Medium (DMEM) is hybridoma tested.
£82.25
Specifications
Content And Storage | 2° to 8°C |
---|---|
Form | Liquid |
Product Type | DMEM Media |
Quantity | 6 x 500 mL |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
15333531
|
Corning™
10-014-CV |
With L-Glutamine, 1g/L Glucose and Sodium Pyruvate |
£82.25
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Growth Promotion and CytotoxicityCorning classical media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications.
Physiochemical Properties
Liquid and powder classical media products are tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology.
Biological Tests
Quality release testing of Corning liquid classical media products assures low levels of endotoxin, mycoplasma, and an absence of contamination.
Endotoxin: Classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) chromogenic assay. For release, each lot of standard liquid classical media must have an endotoxin level of <0.25 EU/mL. Classical media powder products must have an endotoxin level of <0.25 EU/mL when reconstituted to formula weight with low endotoxin water.
Sterility: All Corning liquid classical media products are tested for sterility.
Mycoplasma: Liquid classical media products are tested for mycoplasma using a large-volume method.
Specifications
2° to 8°C | |
Liquid | |
DMEM Media | |
6 x 500 mL |