missing translation for 'onlineSavingsMsg'
Learn More

Cell Culture Tris-Buffered Saline (10X), Corning™

With low endotoxin content

Brand:  Corning Media Tech 46-012-CM

Code : NEW

Additional Details : Weight : 6.98000kg

Product Code. 15363711

  • £324.00 / Pack of 6
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Maintains cell culture media in the physiological pH range
  • Maintains cell tonicity and viability
  • Produced under the highest industry standards to ensure superior results
  • Nontoxic

Specifications

Specifications

10X
Liquid
Sodium Chloride 80g/L, Tris Ultrapure 24.2g/L
7.4
6 x 1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers