missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Ciprofloxacin Hydrochloride
Packaged in plastic bottles
£565.93
Specifications
Content And Storage | 15° to 30°C |
---|---|
Form | Powder |
Product Type | Antibiotic |
Format | Plastic Bottle |
Specifications
15° to 30°C | |
Powder | |
Antibiotic | |
Plastic Bottle |