Learn More
Corning™ Collection Bags
Product Code. p-4668237
Click to view available options
Capacity (Metric):
0.5 L
1 L
10 L
100 L
200 L
Film Description:
EVA
ULDPE
Unit Size:
Each
Product Code. 17283574
Brand: Corning™ 9120042
Collection bags with three ports used for biopharmaceutical applications
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1 L
Tubing Style Heat sealable flexible tubing
Film Description EVA
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site
Quantity Each