missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fetal Bovine Serum, Premium (Embryonic Stem Cell Tested)

Product Code. 15383691
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15383691

Brand: Corning™ 35074CV

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Embryonic Stem (ES) Cell Tested FBS is used by scientists doing embryonic stem cell research. This FBS has been validated for its ability to support colony formation and proper cell morphology with minimal cell differentiation.

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Embryonic Stem Cell Tested FBS is stringently validated for its ability to support colony formation and proper cell morphology while allowing for minimal differentiation. This product is validated with mouse ES cells, but is also suitable for use with Human ES cells.
  • Embryonic Stem Cell Tested FBS is sourced from US herds

Order Info

Shipped in frozen form.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP
Color Amber Yellow to Reddish
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Premium
Endotoxin Concentration <0.084EU/mL
For Use With (Application) Embryonic stem cell research
Hemoglobin Concentration Test and Report
Osmolality Test and Report
pH Test and Report
Protein Content Test and Report
USDA Safety Testing Included USDA Inspected
Show More Show Less

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.