Learn More
Corning™ Flexible Packaging - Tubing Set
Optional tubing sets are available in combination with all single-use bag options
Brand: Corning™ 91-700-00
Additional Details : Weight : 0.05980kg
Description
36„ length 1/4„ I.D. clear chemically resistant, heat sealable, flexible tubingMale Luer and female Luer cap
Female Luer and male Luer cap
Corning media certificates are available on our Resource Library.
Specifications
Tubing Set |