Learn More
Corning™ glutaGRO Supplement
Stabilized dipeptide form of L-Glutamine
Brand: Corning™ 25-015-CI
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- 200 mM solution (100x), with 8.5g/L NaCl
- Does not degrade in liquid media during storage or incubation
- Provides superior results in several applications
- Quantity: 1 x 100mL
Specifications
2° to 30°C | |
glutagro Supplement | |
Plastic Bottle | |
1 x 100 mL |
Liquid | |
Sterile-filtered | |
5 to 6 |