Learn More
Corning™ Non-Gusseted Tank Liners
Designed to fit cylindrical tanks
Brand:  Corning™ 91-300-80
Product Code. 17223594
  • £81.30 / Each
Explore more special offers
Description
Description
This 1090L tank liner is designed to eliminate cross contamination and the need to clean and validate tanks, while extending reusable tank life. Each bag is irradiated and individually packed to ensure sterility. It is ideal for biopharmaceutical applications.
Corning’s portfolio of irradiated tank liners are designed to fit cylindrical tanks. Tank liners are manufactured with low-density polyethylene (ULDPE) and are available in a range of sizes. Features and Benefits
  • Gamma irradiated, individually packaged
  • Free of animal-derived components
  • Eliminate costs associated with the cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate the risks associated with cross contamination
  • Help extend the life of reusable tanks, resulting in lower capital expenditures
  • Wide variety of sizes available
  • Corning media certificates are available on our Resource Library.
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    1090 L
    LDPE
    Biopharmaceutical Applications