Learn More
Corning™ Penicillin-Streptomycin Solution
BIOPHARMA
Effective against Gram-positive and Gram-negative bacteria. Corning™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000μg/mL) in a 100-fold working concentration.
Brand:  Corning™ 30-001-CI
130.35 GBP valid until 2024-12-31
Use promo code "21971" to get your promotional price.
 View more versions of this product
Product Code. 15313671
  • £176.00 / Pack of 6
Explore more special offers
Description
Description
  • Certificate of Analysis available
  • Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-20°C
Solution
Sterile
6 x 100 mL
5000 IU Penicillin, 5000 μg/mL Streptomycin
Antibiotic
Plastic Bottle