missing translation for 'onlineSavingsMsg'
Learn More

Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA

Product Code. 15303661
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
20 L
6 x 1 L
1 x 100 L
1 x 200 L
Packaging:
Bioprocess Container
Bottle
Unit Size:
100 L
20 L
200 L
Pack of 6
This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Product Code. 15303661

Brand: Corning Media Tech 25055LH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Order Info

Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottle
Quantity 1 x 200 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.