missing translation for 'onlineSavingsMsg'
Learn More

Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA

Product Code. 15353651
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
20 L
6 x 1 L
1 x 100 L
1 x 200 L
Packaging:
Bioprocess Container
Bottle
Pakningsstørrelse:
100 L
20 L
200 L
Pack of 6
Denne vare kan ikke returneres. Se returpolicy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Artikelnummer. 15353651

Brand: Corning Media Tech 25055CI

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Ordreinfo

Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.

TRUSTED_SUSTAINABILITY

Tekniske data

Packaging Bottle
Quantity 6 x 100 mL
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.