missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CELLSTACK™ Culture Chambers
Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.
Brand: Corning™ 3313
This item is not returnable.
View return policy
Description
- Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
- Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
- Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
- Sterilized by gamma irradiation
- Leak tested
- Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
- Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation
Order Info
Individually packaged
Certifications
Certified nonpyrogenic.
Compliance
Polystyrene material meets USP Class VI requirements; provides superior optical clarity and structural integrity.
Specifications
21.13 to 32.12 oz. | |
Orange | |
625 to 950 mL | |
Culture Chamber | |
3180 cm² | |
0.2μm | |
Gamma Irradiated |
USP Class VI | |
33mm | |
Polystyrene | |
Yes | |
Tissue Culture-treated | |
Five chambers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction