missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CELLSTACK™ Culture Chambers
GREENER_CHOICEBIOPHARMA

Product Code. 10799494 Shop All Corning Products
Click to view available options
Capacity (Metric):
125 to 190 mL
1250 to 1900 mL
250 to 380 mL
5000 to 7600 mL
625 to 950 mL
Culture Area:
1272 cm²
25,440 cm²
3180 cm²
636 cm²
6360 cm²
Surface Treatment:
CellBIND
Tissue Culture-treated
Ultra-low Attachment
Quantity:
10 chambers
40 chambers
Five chambers
One chamber
Two chambers
Unit Size:
Pack of 2
Pack of 5
Pack of 6
Pack of 8
This item is not returnable. View return policy

Product Code. 10799494

Brand: Corning™ 3270

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.

  • Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
  • Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
  • Sterilized by gamma irradiation
  • Leak tested

  • Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
  • Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation

Order Info

Individually packaged

Certifications

Certified nonpyrogenic.

Compliance

Polystyrene material meets USP Class VI requirements; provides superior optical clarity and structural integrity.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 42.26 to 64.24 oz.
Certifications/Compliance USP Class VI
Closure Color Orange
Diameter (Metric) Closure 33mm
Capacity (Metric) 1250 to 1900 mL
Material Polystyrene
Product Type Culture Chamber
Graduated Yes
Culture Area 6360 cm²
Surface Treatment Tissue Culture-treated
Quantity 10 chambers
Sterilization Method Gamma Irradiated
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.