missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen I, Rat
BIOPHARMA

Product Code. p-4696757 Shop All Corning Products
Click to view available options
Quantity:
1000 mg
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10727531

Brand: Corning™ 356236

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 3 to 4 mg/mL
For Use With (Application) Cell Attachment and Proliferation, Cell-specific Morphology and Function
Formulation Liquid in 0.02N acetic acid
Quantity 1000 mg
Source Rat Tail Tendon
Product Type Collagen, Rat Tail Type I
Content And Storage 2°C to 8°C
Purity or Quality Grade >90% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.