Learn More
Corning™ Color-Coded Cap Inserts
For use with Corning cryogenic vials except 03-374-6
Brand:  Corning™ 2015
Code : 66
Additional Details : Weight : 0.08820kg
 View more versions of this product
Product Code. 10317532
  • £40.04 / Pack of 500
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Cap inserts provide color coding for easy sample identification.
  • Inserts are packaged in resealable bags.
  • Nonsterile
  • Cap inserts fit all Corning™ cryogenic vials.
Specifications
Specifications
White
Polypropylene
500
Polypropylene
White
Non-sterile
50