Learn More
Corning™ Color-Coded Cap Inserts
For use with Corning cryogenic vials except 03-374-6
Marque: Corning™ 430499
Code : 66
Informations supplémentaires : Weight : 0.09000kg
Description
- Cap inserts provide color coding for easy sample identification.
- Inserts are packaged in resealable bags.
- Nonsterile
- Cap inserts fit all Corning™ cryogenic vials.
Spécification
Assorted | |
Polypropylene | |
500 | |
Polypropylene |
Assorted | |
Non-sterile | |
50 |