Learn More
Corning™ Costar™ Tip Loading System Refills
System consists of a one-piece base which attaches to a “magazine” consisting of 10 layers of 96 tips
Brand: Corning™ 4785
Code : 66
Additional Details : Weight : 1.00000kg
Specifications
1 to 200 μL | |
Yellow | |
Rack | |
Costar |