missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cover Glasses

Product Code. 10602184 Shop All Corning Products
Click to view available options
Dimensions (L x W):
22 x 22 mm
Shape:
Square
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10602184

Brand: Corning™ 286522

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fine optical glass developed specifically for cover glasses

  • Free of stones, blisters, bubbles and striae
  • High resistance to surface attack, weather

Order Info

  • Packaged in lint-free reclosable plastic boxes for complete protection.
  • Supplied in packs of 1 oz.

TRUSTED_SUSTAINABILITY

Specifications

Shape Square
Dimensions (L x W) 22 x 22 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.