Learn More
Corning™ CrystalEX384-Well Flat Bottom Protein Crystallization Microplate
GREENER_CHOICE
The only microplate designed for full automation in crystal screening
Brand:  Corning™ 3775
Product Code. 10285862
  • £1165.00 / Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
96-well Microplates
  • Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
  • Conical bottom protein wells allow improved centering of the protein drop.
  • Compatible with manual pipettors and automation
  • Novel merged well design provides efficient vapor space for protein crystallization.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Features 192 reservoir wells and 192 corresponding protein wells
  • Flat bottom protein wells are optimized for imaging of crystals.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Advanced Polymer
Non-sterile
Flat Bottom (F-well)
50/Cs.
50/Cs.
Without Lid
Clear
384
Non-treated
192 reservoir wells and 192 corresponding protein wells
10/Pk.
2.3 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products