missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CrystalEX™ 96-Well Protein Crystallization Microplates
Designed for sitting-drop vapor diffusion high throughput protein crystallization
£639.81 - £640.00
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
No. of Wells | 96 |
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
10039661
|
Corning™
3773 |
Non-treated |
£639.81
Pack of 50 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
10124162
|
Corning™
3785 |
High Binding |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
96-well Microplates
- Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
- Conical bottom protein wells allow improved centering of the protein drop.
- Compatible with manual pipettors and automation
- Novel merged well design provides efficient vapor space for protein crystallization.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
- Meets ANSI/SBS standard footprint dimensions for microplates
- Ideal for fully automated crystal screening
- Features 192 reservoir wells and 192 corresponding protein wells
- Flat bottom protein wells are optimized for imaging of crystals.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
Specifications
Standard | |
8 x 12 | |
96 | |
96 flat-bottom reservoir wells and 96 conical-bottom protein wells | |
50/Cs. |
Without Lid | |
Clear | |
Non-sterile | |
50/Cs. | |
3.1 mm |