Learn More
Corning™ Untreated Culture Dishes
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Brand: Corning™ 430597
Code : 67
Additional Details : Weight : 4.05000kg
Description
- Manufactured from optically clear virgin polystyrene
- Not treated for applications where cell attachment is not desired
- Not treated for applications where cell attachment is not desired
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Sterile
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431111, Square dishes with interior bottom dimensions of 224 x 224 mm.
Specifications
Clear | |
Polystyrene | |
45 mL (Well) | |
148 cm2 | |
Polystyrene | |
Sterile | |
Cell Culture Petri Dish | |
60/Cs. | |
25 mm |
Yes | |
Flat | |
Clear | |
Single-use | |
Round | |
Non-treated | |
Lid | |
5/Pk. | |
150 mm |