missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips

Product Code. p-4110854 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 100 μL
1 to 20 μL
1 to 200 μL
Unit Size:
Pack of 3840
Pack of 960

Product Code. 11759325

Brand: Corning™ 4135

Only null left
Add to basket
Add to basket

Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures

Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Filtered
Sterility Sterile
Color Natural
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Low Retention, Round
Packaging Rack
Graduated Yes
Product Line DeckWorks
Material HDPE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.