Learn More
Corning™ DeckWorks™ Reload Systems with Standard Tips
Packaged in transparent trays for easy tip identification. Corning™ Deckworks™ Reload Systems with Standard Tips provide convenient and economical reloading of durable hinged racks
Brand: Corning™ 4129
42.74 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
Corning DeckWorks reload systems are available in 10 μL, 200 μL, 300 μL, and 1,000 μL sizes and allow for convenient and economical reloading of durable Corning DeckWorks hinged racks. DeckWorks reload decks are packaged in transparent trays for easy tip identification, requires no transfer devices, saves space, and minimizes packaging waste. Although the pipet tips are made from virgin medical-grade resins, the reload decks and rack bases are manufactured from 100% recycled polypropylene. Nonsterile decks and racks are steam autoclavable.
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips is eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins, and/or peptides.
Specifications
100 to 1000 μL | |
Natural | |
Rack | |
Reload system | |
DeckWorks |
Non-sterile | |
Autoclavable | |
Round | |
Yes | |
Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.