Learn More
Corning™ Digital Hotplate and Stirrer Accessory Kit
GREENER_CHOICE
For Use With: PC-400D, PC-420D, PC-600D, PC-620D Digital Display Hotplates and Stirring Hotplates
Brand:  Corning™ 6795KIT
Product Code. 10451138
  • £124.31 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This accessory kit is designed for all Corning digital display Hotplates and stirring Hotplates (Models PC-400D, PC-420D, PC-600D, PC-620D).
This Corning™ kit Cat. No. 6795KIT includes:
  • Cat. No. 6795PR, external temperature controller
  • Cat. No 440129, vertical support rod. Supplied as two 9 inch (22.9 cm) support rods that can be screwed together (18 inches)
  • Cat. No. 6970SR, stir bar retriever
Available for purchase separately and not included in the kit:
  • Cat. No. 440140, boss head clamp
  • Cat. No. 440141, holding rod
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.8 cm
Temperature Controller (6795PR), A Two Piece, 0.8 cm Diameter, 45.7 cm L Support Rod (440129) and Magnetic Stir Bar Retriever (6970SR)
Hot Plate Kit
PC-400D, PC-420D, PC-600D, PC-620D Digital Display Hotplates and Stirring Hotplates
45.7 cm