Learn More
Corning™ Disposable Glass Pasteur Pipets, Non-Sterile, Unplugged
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
£54.85 - £102.68
Specifications
Aspirator Plug | No |
---|---|
Material | Soda Lime Glass |
Includes | 1 in. Constriction below the top |
Tip Style | Round |
Type | Unplugged Pasteur Pipet |
Product Code | Brand | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
10653004
|
Corning™
7095B-NMR |
33.02 cm | 0.65 cm |
£102.68
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
10343663
|
Corning™
7095B-5X |
14.6 cm | 0.65 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10528603
|
Corning™
7095B 9PK |
22.86 cm | 0.55 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid nonvolumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
'B' series pipets are soda lime glass. 'D' series are borosilicate glass.
Specifications
No | |
1 in. Constriction below the top | |
Unplugged Pasteur Pipet |
Soda Lime Glass | |
Round | |
Single-use |