missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Easy Grip Disposable Polystyrene Sterile Bottles
Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
Brand: Corning™ 431175
81.73 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Description
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Corning low profile style has easy grip sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Nonpyrogenic
Specifications
Single-use | |
Sterile | |
Orange HDPE plug seal | |
24/Cs. | |
24 Case | |
150 mL |
Polystyrene | |
Applications: Storage of media, buffers and other aqueous solutions | |
4.5 cm | |
2/Pk. | |
Graduated Easy Grip Storage Bottle |