missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Easy Grip Disposable Polystyrene Sterile Bottles
GREENER_CHOICEBIOPHARMA

Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Brand:  Corning™ 430281

87.36 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 10738212

  • £103.00 / Pack of 24
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Corning low profile style has easy grip sides that facilitate handling
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Sterile
Orange HDPE plug seal
24/Cs.
24 Case
250 mL
Polystyrene
Applications: Storage of media, buffers and other aqueous solutions
4.5 cm
2/Pk.
Graduated Easy Grip Storage Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Easy Grip Disposable Polystyrene Sterile Bottles > Volume: 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.