missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Easy Grip Disposable Polystyrene Sterile Bottles

Shop alle Corning produkter


Klik for at se tilgængelige muligheder
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
Pakningsstørrelse:
Pack of 24
Beskrivelse
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Corning low profile style has easy grip sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Nonpyrogenic
Tekniske data
Tekniske data
| Capacity (Metric) | 150 mL |
| For Use With (Application) | Applications: Storage of media, buffers and other aqueous solutions |
| Includes | Orange HDPE plug seal |
| Material | Polystyrene |
| No. per Case | 24/Cs. |
| No. per Pack | 2/Pk. |
| Quantity | 24 Case |
| Sterility | Sterile |
| Disposable | Single-use |
| Mouth | 4.5 cm |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion